Learn More
Thermo Scientific Chemicals D-Glutamic acid, 99+%
CAS: 6893-26-1 | C5H9NO4 | 147.13 g/mol
$91.92 - $856.55
Identifiants chimiques
| CAS | 6893-26-1 |
|---|---|
| Molecular Formula | C5H9NO4 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD00063112 |
| InChI Key | WHUUTDBJXJRKMK-GSVOUGTGSA-N |
| Synonym | d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid |
| PubChem CID | 23327 |
| ChEBI | CHEBI:15966 |
| IUPAC Name | (2R)-2-aminopentanedioic acid |
| SMILES | C(CC(=O)O)C(C(=O)O)N |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAA1419106
|
Thermo Scientific Chemicals
A1419106 |
5 g |
chaque for $91.92
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA1419114
|
Thermo Scientific Chemicals
A1419114 |
25 g |
chaque for $271.59
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA1419122
|
Thermo Scientific Chemicals
A1419122 |
100 g |
chaque for $856.55
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.
Solubility
Soluble in water (8.88 mg/mL) at 25°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Identifiants chimiques
| 6893-26-1 | |
| 147.13 | |
| WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
| 23327 | |
| (2R)-2-aminopentanedioic acid |
| C5H9NO4 | |
| MFCD00063112 | |
| d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
| CHEBI:15966 | |
| C(CC(=O)O)C(C(=O)O)N |
Spécifications
| 199°C (decomposition) | |
| C5H9NO4 | |
| 1723800 | |
| d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
| WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
| (2R)-2-aminopentanedioic acid | |
| 147.13 | |
| CHEBI:15966 | |
| 147.13 | |
| D-Glutamic acid |
| 6893-26-1 | |
| MFCD00063112 | |
| 14,4469 | |
| Soluble in water (8.88mg/mL) at 25°C. | |
| C(CC(=O)O)C(C(=O)O)N | |
| 5 g | |
| 23327 | |
| −30° (c=10 in 2N HCl) | |
| ≥99% |
Sécurité et manipulation
EINECSNumber : 230-000-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only