Learn More
Cyclopentanol, 99%
CAS: 96-41-3 | C5H10O | 86.134 g/mol
$57.21 - $699.05
Identifiants chimiques
| CAS | 96-41-3 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.134 |
| MDL Number | MFCD00001363 |
| InChI Key | XCIXKGXIYUWCLL-UHFFFAOYSA-N |
| Synonym | hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol |
| PubChem CID | 7298 |
| ChEBI | CHEBI:16133 |
| IUPAC Name | cyclopentanol |
| SMILES | C1CCC(C1)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAA15249AE
|
Thermo Scientific Chemicals
A15249AE |
100 mL |
chaque for $57.21
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA15249AP
|
Thermo Scientific Chemicals
A15249AP |
500 mL |
chaque for $206.53
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAA152490F
|
Thermo Scientific Chemicals
A152490F |
2500 mL |
chaque for $699.05
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopentanol is a versatile compound used as a building block, intermediate and solvent.
Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Identifiants chimiques
| 96-41-3 | |
| 86.134 | |
| XCIXKGXIYUWCLL-UHFFFAOYSA-N | |
| 7298 | |
| cyclopentanol |
| C5H10O | |
| MFCD00001363 | |
| hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol | |
| CHEBI:16133 | |
| C1CCC(C1)O |
Spécifications
| 96-41-3 | |
| 0.95 | |
| 51°C (123°F) | |
| C5H10O | |
| MFCD00001363 | |
| UN2244 | |
| 14,2742 | |
| Slightly soluble in water. Also soluble in ether,alcohols and acetone. | |
| C1CCC(C1)O | |
| 86.134 | |
| CHEBI:16133 | |
| 99% |
| -19°C | |
| 139°C to 141°C | |
| Unpleasant | |
| 1.453 | |
| 100 mL | |
| 1900556 | |
| hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol | |
| XCIXKGXIYUWCLL-UHFFFAOYSA-N | |
| cyclopentanol | |
| 7298 | |
| 86.13 | |
| Cyclopentanol |
Sécurité et manipulation
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P243-P261-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P374-P380-P501c
H226-H302-H310-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANOL
missing translation for 'einecsNumber' : 202-504-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only