Learn More
Cyclopentane, 97%, extra pure
Cyclopentane, 97%, C5H10, CAS Number-287-92-3, ciclopentano, cyclopentan, cyclopentanes, cyclopentyl group, hsdb 62, hydrocarbons, cyclic c5 and c6, pentamethylene, t86pb90rnu, unii-t86pb90rnu, zyklopentan, 1L, 96% min. (GC), 05, 19, 50.0 deg.C, CHEBI:23492, White, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol
Supplier: Thermo Scientific Chemicals 111480025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Cyclopentane | |
| -94.0°C | |
| 0.7500g/mL | |
| −7°C | |
| 96% min. (GC) | |
| C5H10 | |
| C5H10 | |
| 2.5 L | |
| 0.01 % max. | |
| 15, 2736 | |
| Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents | |
| C1CCCC1 | |
| 70.15 | |
| 0.44 mPa.s (20°C) | |
| 70.15 | |
| Extra Pure |
| 287-92-3 | |
| White | |
| 50.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4050 to 1.4070 | |
| MFCD00001356 | |
| 05, 19 | |
| 0.75 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| cyclopentane | |
| 9253 | |
| CHEBI:23492 | |
| 97% | |
| Powder |
Identifiants chimiques
| 287-92-3 | |
| 70.15 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| 9253 | |
| cyclopentane |
| C5H10 | |
| MFCD00001356 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| CHEBI:23492 | |
| C1CCCC1 |
Sécurité et manipulation
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-016-6
RUO – Research Use Only