missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone, Reagents
Cyclohexanone, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2193600500C
Spécifications
| Cyclohexanone | |
| 108-94-1 | |
| 154°C | |
| Glass Bottle | |
| 0.9478 | |
| Pimelic Ketone, Ketohexamethylene | |
| O=C1CCCCC1 | |
| 98.15 |
| -28.9°C | |
| Colorless or Pale Yellow | |
| 500 mL | |
| Liquid | |
| C6H10O | |
| JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
| cyclohexanone | |
| Reagent |
Identifiants chimiques
| 108-94-1 | |
| 98.15 | |
| Pimelic Ketone, Ketohexamethylene | |
| O=C1CCCCC1 |
| C6H10O | |
| JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
| cyclohexanone |