Learn More
Cyanoacetic acid, 97%
CAS: 372-09-8 | C3H3NO2 | 85.06 g/mol
Supplier: Thermo Scientific Chemicals 110670010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Cyanoacetic acid | |
| 65.0°C to 67.0°C | |
| 108.0°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| NCCH2CO2H | |
| 02,583 | |
| 15,2679 | |
| Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene and chloroform | |
| C(C#N)C(=O)O | |
| 85.06 | |
| CHEBI:51889 | |
| 97% |
| 372-09-8 | |
| White to Beige | |
| 107°C | |
| 97% | |
| C3H3NO2 | |
| 1 kg | |
| 11,145; 13,171 | |
| cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova | |
| MLIREBYILWEBDM-UHFFFAOYSA-N | |
| 2-cyanoacetic acid | |
| 9740 | |
| 85.06 | |
| Adhering Crystalline Solid |
Identifiants chimiques
| 372-09-8 | |
| 85.06 | |
| cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova | |
| CHEBI:51889 | |
| C(C#N)C(=O)O |
| C3H3NO2 | |
| MLIREBYILWEBDM-UHFFFAOYSA-N | |
| 9740 | |
| 2-cyanoacetic acid |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-743-9
RUO – Research Use Only