missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ One SFM

Numéro de catalogue. A5757204
Modifier l'affichage
Click to view available options
Quantité:
1 L
5 L
10 L
Format:
Bottle
BPC
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Format
A5757204 10 L BPC
A5757201 1 L Bottle
A5757202 1 L BPC
A5757203 5 L BPC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. A5757204 Fournisseur Gibco™ Code fournisseur A5757204

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies.

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies. This medium improves cell count and early memory phenotype throughout all stages of expansion.

Benefits:

  • Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
  • Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
  • Expands both healthy and disease state donors
  • Supports efficient viral and non-viral editing with downstream expansion
  • Mitigates risk of contaminants and variability associated with animal-sourced materials
  • Supports scale-up via larger catalog pack sizes

Features:

  • Animal-origin-free (AOF) formulation
  • Serum-free—designed without the need for serum or serum replacement
  • No separate components—no need to mix basal medium with supplement to obtain complete medium
  • Yields high density cultures
  • Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD28 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
  • Available in 1 L bottle, 1 L bag, 5 L bag, and 10 L bag options

Increased efficiency and consistent performance:

CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.

Simplify your transition into clinical research:

The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)

Spécifications

Classification Animal Origin-free, Serum-free, cGMP
Forme Liquid
Taux de sérum Serum-free
Stérilité Sterile-filtered
Méthode de stérilisation 0.2 μm Sterile-filtered
Avec additifs None
Sans additif No Glutamine, No Phenol Red
Gamme de produit CTS OpTmizer
Quantité 10 L
Format BPC
Contenu et stockage Store at 2-8°C and protect from light.
Type de culture Suspension Cell Culture, Dynamic Bioreactors
Afficher plus de résultats Afficher moins de résultats

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.