Learn More
Gibco™ CTS™ N-2 Supplement
Description
CTS N-2 Supplement is a chemically defined, serum-free supplement that supports the growth and viability of pluripotent stem cells (PSCs) during differentiation and a variety of PSC-derived cell types. It offers equivalent performance to the research version and contains the same optimized concentrations of insulin, holo-transferrin, progesterone, putrescine, and selenite. The CTS version meets higher quality standards, is GMP manufactured, and includes regulatory documentation, making it suitable for cell therapy manufacturing.
CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Accompanied by documentation to support your regulatory filing
- Access to our Drug Master File (DMF) or Regulatory Support File (RSF)
- Traceability documentation: Certificate of Origin, Certificate of Analysis, and Safety Data Sheet
Consider using CTS N-2 Supplement with CTS Neurobasal Medium or other CTS cell culture media to support your cell therapy production.
Specifications
Specifications
| Cell Type | Mammalian Cells |
| Classification | Xeno-free |
| Concentration | 100 X |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21 CFR820 |
| Product Line | CTS |
| Show More |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.