missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Crustacean Cardioactive Peptide
For cellular and molecular biology applications
$974.45 - $974.45
Spécifications
| Purification Method | HPLC |
|---|---|
| Common Name | CCAP |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 957.10 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAJ66453MA
|
Thermo Scientific Chemicals
J66453MA |
10 mg |
chaque for $974.45
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| HPLC | |
| -30°C to -10°C | |
| 957.10 | |
| RUO - research use only |
| CCAP | |
| Solid | |
| Crustacean Cardioactive Peptide | |
| Pro-Phe-Cys-Asn-Ala-Phe-Thr-Gly-Cys (Disulfide bridge: Cys3-Cys9) |
RUO – Research Use Only