missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red TS, 0.04% (w/v), Ricca Chemical

Chemical Identifiers

CAS 62625-29-0 , 7732-18-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
24504
View Documents Promotion Details
N/A 120 mL Natural Poly Bottle N/A
Only null left
 
245016
View Documents Promotion Details
N/A 500 mL Natural Poly Bottle N/A
Only null left
 
R24500001A
View Documents Promotion Details
N/A 1 L Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-29-0, 7732-18-5

Specifications

62625-29-0 , 7732-18-5
100.0, 0.04
0.04% W/V
Aqueous Solution
Liquid
98.0, 0.04
0.04% (w/v)
Indicator
Red
Cresol Red, Test Solution
Videos
SDS
Documents

Documents