CAS RN 62625-29-0
IUPAC Name:
sodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Synonyms:
sodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzenolate
Molecular Weight (g/mol):
404.41
Molecular Formula:
C21H17NaO5S
InChi Key:
FMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES:
[Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
1
–
9
of
9
results
1
Thermo Scientific Chemicals Cresol Red sodium salt, 0.04% w/v aq. soln.
CAS: 62625-29-0 Molecular Formula: C21H17O5S MDL Number: MFCD00001618
2
Promotions Available
Thermo Scientific Chemicals Cresol Red sodium salt
Thermo Scientific Chemicals Cresol Red sodium salt
CAS: 62625-29-0 MDL Number: MFCD00001618 Synonym: o-Cresolsulfonephthalein sodium salt
3
Promotions Available
Thermo Scientific Chemicals Cresol Red, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals Cresol Red, sodium salt, pure, water soluble, indicator
CAS: 62625-29-0 Molecular Formula: C21H17NaO5S MDL Number: MFCD00001618
4
Cresol Red Sodium Salt, Reagent Grade, LabChem™
CAS: 62625-29-0
5
Cresol Red Sodium Salt, Ricca Chemical
CAS: 62625-29-0
6
Cresol Red (0.04% in Water) [for pH Determination], TCI America™
CAS: 62625-29-0 MDL Number: MFCD00001618
7
Cresol Red Sodium Salt, TCI America™
CAS: 62625-29-0 Molecular Formula: C21H18O5S MDL Number: MFCD00001618 Synonym: Sodium Cresol Red
8
Cresol Red TS, 0.04% (w/v), Ricca Chemical
CAS: 62625-29-0