missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red TS, 0.04% (w/v), Ricca Chemical

Catalog No. 24504
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
24504 120 mL Natural Poly Bottle
245016 500 mL Natural Poly Bottle
R24500001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24504 Supplier Ricca Chemical Company Supplier No. 24504

Chemical Identifiers

CAS 62625-29-0 , 7732-18-5

Specifications

CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Concentration 0.04% (w/v)
Percent Purity 0.04% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Red
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Cresol Red, Test Solution
Show More Show Less