missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol Red TS, 0.04% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 245016
| Quantity | 500 mL |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| 62625-29-0, 7732-18-5 |
Specifications
| Cresol Red | |
| 62625-29-0 , 7732-18-5 | |
| 100.0, 0.04 | |
| 0.04% W/V | |
| Natural Poly Bottle | |
| Red | |
| Liquid |
| Test Solution | |
| 98.0, 0.04 | |
| 0.04% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 500 mL |