Learn More
Scientific Industries Inc COUNTER BASIC PILL AND BALANCE DFS Item
Supplier:  Scientific Industries Inc DRX4C2320
Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing. Equipped with a basic pill counting feature this balance is an ideal entry level pill counter for any pharmacy. This unit is great for users that like the features of the DRX-4C but have no bar-coded drugs. Capacity is 320g x 0.001g with backlit LCD.
Catalog No. 501139341
Only null left
Explore available promotions