Learn More
Corning™ RPMI 1640 Medium (Mod.)
Catalog No. MT50020PC
Change view
Click to view available options
Quantity:
10000 mL
50000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT50020PC 50000 mL
MT50020PB 10000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT50020PC Supplier Corning™ Supplier No. 50020PC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol.  Examples of cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.  RPMI 1640 is also used for suspension cultures or monolayer human leukemic cells.  This formulation contains L-glutamine and does not contain sodium bicarbonate.
Specifications
Without Additives No Sodium Bicarbonate
Quantity 50000 mL