missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99.9%, (trace metal basis), shot 1-10 mm

Catalog No. AC207695000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207695000 500 g Glass bottle
AC207691000 100 g Glass bottle
AC207690025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC207695000 Supplier Thermo Scientific Chemicals Supplier No. 207695000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Quantity 500 g
Packaging Glass bottle
Form Granular
Assay Percent Range 99.9%
Linear Formula Cu
Fieser 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Copper
Show More Show Less

RUO – Research Use Only