Learn More
Mopec™ Control Slides, Special Stains, Liver Tissue with Fungus (Periodic Acid Schiff)
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
499,73 $ - 2 614,07 $
Spécifications
Type Control Slide
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantité  
22-457-727
Afficher les documents
Mopec™
SC82410
10 Unstained, 1 Stained Slide N/A
Il en reste null
22-457-788
Afficher les documents
Mopec™
SC82420
20 Unstained, 2 Stained Slides N/A
Il en reste null
22-457-526
Afficher les documents
Mopec™
SC824100
98 Unstained, 2 Stained Slides N/A
Il en reste null
Description
Description
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 20 contains 20 unstained slides and 2 stained slides
  • Case of 100 contains 98 unstained slides and 2 stained slides
Spécifications
Spécifications
Control Slide
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'