missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polynit Heatseal Wipes
GREENER_CHOICE

Laundered and packaged in an ISO Class 4 cleanroom. Contec™ Polynit Heatseal Wipes are available with Particle Attraction Technology (PAT).

$943.96

Spécifications

Material Polyester
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Product Type Packaging Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Dimensions (L x W) Product Type Packaging Quantity Prix Quantity & Availability  
19170950
GREENER_CHOICE
Afficher les documents Détails de la promotion
Contec™
LWPS0027
9 x 9 in. (22.86 x 22.86 cm) Cleanroom Wipe 1200 Wipes/Bag
caisse de 1200 chaque for $943.96
Il en reste null
 
Description

Description

 

  • 100% textured polyester wipe with nonabrasive, heat-sealed edge
  • Chemical- and abrasion-resistant, with good solvent sorbency
  • Extremely low in both particles, fibers, and nonvolatile residues
  • Specially treated to enhance particle attraction and retention
  • Attracts 35 times more particles than traditional wipes
  • Retains 95% of particles captured, so there is less chance of re-contamination

 

Wipes with Particle Attraction Technology (PAT)

  • 19-170-930: 9 x 9 in.
  • 19-170-931: 12 x 12 in.
  • 19-170-932: 4 x 4 in.
  • 19-170-933: 9 x 9 in.

 

Applications:

Ideal for general-purpose cleaning and product wipe down.

Spécifications

Spécifications

Polyester
Videos
FDS
Documents

Documents