missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C (30,300 ppm TDS as NaCl), Ricca Chemical
$125.10 - $489.17
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2248-16
|
Ricca Chemical Company
224816 |
500 mL | Natural Poly Bottle |
Each for $125.10
|
|
||||
|
2248-32
|
Ricca Chemical Company
RI224832 |
1 L | Natural Poly Bottle |
Each for $204.60
|
|
||||
|
2248-1
|
Ricca Chemical Company
RI22481 |
4 L | Natural Poly Bottle |
Each for $489.17
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Specifications
| Natural Poly Bottle | |
| 30,300ppm as NaCl | |
| ClNa | |
| Colorless | |
| Conductivity Standard | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 |
| 7647-14-5,7732-18-5 | |
| Liquid | |
| MFCD00003477 | |
| Approximately 0°C | |
| Conductivity Standard | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 | |
| Conductivity/TDS Standard |