missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C (30,300 ppm TDS as NaCl), Ricca Chemical

Catalog No. 224816
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
224816 500 mL Natural Poly Bottle
224832 1 L Natural Poly Bottle
22481 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 224816 Supplier Ricca Chemical Company Supplier No. 224816
Only null left

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 30,300ppm as NaCl
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Conductivity/TDS Standard