missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 50,000 μS/cm (50 mS/cm) at 25°C (30,300 ppm TDS as NaCl), Ricca Chemical

Supplier:  Ricca Chemical Company RI22481

 View more versions of this product

Catalog No. 2248-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Conductivity/TDS Standard
7647-14-5,7732-18-5
Liquid
MFCD00003477
Approximately 0°C
Conductivity Standard
[Na+].[Cl-]
58.44
5234
Natural Poly Bottle
30,300ppm as NaCl
ClNa
Colorless
Conductivity Standard
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
4 L
CHEBI:26710

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions