missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 3000 μS/cm at 25°C (1534 ppm TDS as NaCl), Ricca Chemical
$136.77 - $439.67
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
|---|---|
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5 , 7732-18-5 |
| CAS Min % | 98.0,0.15 |
| Concentration or Composition Notes | 1534ppm as NaCl |