missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 3000 μS/cm at 25°C (1534 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company 22445016
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5,7732-18-5 | |
| 1534ppm as NaCl | |
| Liquid | |
| 100°C | |
| Conductivity Standard |
| Natural Poly Bottle | |
| 98.0,0.15 | |
| 100.0,0.16 | |
| Colorless | |
| Conductivity Standard | |
| 500 mL |