missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 3000 μS/cm at 25°C (1534 ppm TDS as NaCl), Ricca Chemical

Catalog No. 22445032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22445032 1 L
22445016 500 mL
2244501 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22445032 Supplier Ricca Chemical Company Supplier No. 2244.5032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 1534ppm as NaCl
CAS Min % 98.0, 0.15
CAS Max % 100.0, 0.16
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Show More Show Less