missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 2000 μS/cm at 25°C (1000 ppm TDS as NaCl), Ricca Chemical
$106.09 - $459.87
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2244-16
|
Ricca Chemical Company
224416 |
500 mL | Natural Poly Bottle |
Each for $106.09
|
|
||||
|
2244-32
|
Ricca Chemical Company
224432 |
1 L | Natural Poly Bottle |
Each for $192.43
|
|
||||
|
2244-1
|
Ricca Chemical Company
22441 |
4 L | Natural Poly Bottle |
Each for $459.87
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Specifications
| Natural Poly Bottle | |
| 1000ppm as NaCl | |
| ClNa | |
| Colorless | |
| Conductivity Standard | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 |