missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 2000 μS/cm at 25°C (1000 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company 22441
Specifications
| Conductivity/TDS Standard | |
| 7647-14-5 , 7732-18-5 | |
| Liquid | |
| MFCD00003477 | |
| 100°C | |
| Conductivity Standard | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 |
| Natural Poly Bottle | |
| 1000ppm as NaCl | |
| ClNa | |
| Colorless | |
| Conductivity Standard | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 4 L | |
| CHEBI:26710 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |