Learn More
Eisco™ Clear Reagent Bottle with Screw Cap
High quality, borosilicate glass flask. Clear Reagent Bottle with Screw Cap and pouring ring for drip free pouring and clean, safe working.
$29.07 - $163.93
Specifications
Color Clear
Material Borosilicate Glass
Quantity 1
Sterility Non-sterile
Closure Included Yes
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S27982
View Documents Promotion Details
Eisco™
CH0164B
250 mL
Each for $39.96
Only null left
S27983
View Documents Promotion Details
Eisco™
CH0164C
500 mL
Each for $48.50
Only null left
S27984
View Documents Promotion Details
Eisco™
CH0164D
1000 mL
Each for $101.18
Only null left
S27985
View Documents Promotion Details
Eisco™
CH0164E
2000 mL
Each for $163.93
Only null left
S27980
View Documents Promotion Details
Eisco™
CH0164A
100 mL
Each for $29.07
Only null left
Description
Description
  • Reagent bottle made high quality, transparent Borosilicate 3.3 Glass
  • Ideal for storing chemicals in liquid or powder form
  • Includes a blue, GL45 screw cap to ensure an airtight seal
  • Features a drip free pouring ring. Both screw cap and pouring ring are autoclavable
  • Bottle features white graduation lines
  • Autoclavable, with superior chemical durability
  • Low temperature gradient- can withstand high temperatures and thermal shocks
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Specifications
Specifications
Clear
1
Yes
Yes
Cylindrical
Chemicals in liquid or powder form
GL45
Screw
Borosilicate Glass
Non-sterile
Reusable
Narrow
Reagent Bottle
Unlabeled
3 cm
Autoclavable