missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Clear Reagent Bottle with Screw Cap

High quality, borosilicate glass flask. Clear Reagent Bottle with Screw Cap and pouring ring for drip free pouring and clean, safe working.

Supplier:  Eisco™ CH0164A

 View more versions of this product

Catalog No. S27980


Only null left

Description

Description

  • Reagent bottle made high quality, transparent Borosilicate 3.3 Glass
  • Ideal for storing chemicals in liquid or powder form
  • Includes a blue, GL45 screw cap to ensure an airtight seal
  • Features a drip free pouring ring. Both screw cap and pouring ring are autoclavable
  • Bottle features white graduation lines
  • Autoclavable, with superior chemical durability
  • Low temperature gradient- can withstand high temperatures and thermal shocks
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Specifications

Chemical Identifiers

Clear
1
Yes
Yes
Cylindrical
Chemicals in liquid or powder form
GL45
Screw
Borosilicate Glass
Non-sterile
Reusable
Narrow
Reagent Bottle
Unlabeled
3 cm
Autoclavable

Specifications

100 mL
3.38 oz.
Borosilicate Glass
Non-sterile
Yes
Yes
Cylindrical
Reagent Bottle
4 in.
GL45
Screw
100 mL
Clear
1
10 mL
Reusable
Narrow
10 cm
Chemicals in liquid or powder form
Unlabeled
3 cm
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions