missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Clear Reagent Bottle with Screw Cap
High quality, borosilicate glass flask. Clear Reagent Bottle with Screw Cap and pouring ring for drip free pouring and clean, safe working.
Supplier: Eisco™ CH0164C
Description
- Reagent bottle made high quality, transparent Borosilicate 3.3 Glass
- Ideal for storing chemicals in liquid or powder form
- Includes a blue, GL45 screw cap to ensure an airtight seal
- Features a drip free pouring ring. Both screw cap and pouring ring are autoclavable
- Bottle features white graduation lines
- Autoclavable, with superior chemical durability
- Low temperature gradient- can withstand high temperatures and thermal shocks
- Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Chemical Identifiers
| Clear | |
| 1 | |
| Yes | |
| Yes | |
| Cylindrical | |
| Chemicals in liquid or powder form | |
| GL45 | |
| Screw |
| Borosilicate Glass | |
| Non-sterile | |
| Reusable | |
| Narrow | |
| Reagent Bottle | |
| Unlabeled | |
| 3 cm | |
| Autoclavable |
Specifications
| 500 mL | |
| 17 oz. | |
| Borosilicate Glass | |
| Non-sterile | |
| Yes | |
| Yes | |
| Cylindrical | |
| Reagent Bottle | |
| 5 in. | |
| GL45 | |
| Screw |
| 500 mL | |
| Clear | |
| 1 | |
| 50 mL | |
| Reusable | |
| Narrow | |
| 12.7 cm | |
| Chemicals in liquid or powder form | |
| Unlabeled | |
| 3 cm | |
| Autoclavable |