missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Clear Reagent Bottle with Screw Cap

Catalog No. S27982
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27982 250 mL
S27983 500 mL
S27984 1000 mL
S27985 2000 mL
S27980 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S27982 Supplier Eisco™ Supplier No. CH0164B

High quality, borosilicate glass flask. Clear Reagent Bottle with Screw Cap and pouring ring for drip free pouring and clean, safe working.

  • Reagent bottle made high quality, transparent Borosilicate 3.3 Glass
  • Ideal for storing chemicals in liquid or powder form
  • Includes a blue, GL45 screw cap to ensure an airtight seal
  • Features a drip free pouring ring. Both screw cap and pouring ring are autoclavable
  • Bottle features white graduation lines
  • Autoclavable, with superior chemical durability
  • Low temperature gradient- can withstand high temperatures and thermal shocks
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use

Specifications

Capacity 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Material Borosilicate Glass
Quantity 1
Sterility Non-sterile
Graduations 25 mL
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Narrow
Shape Cylindrical
Height (Metric) 10 cm
Product Type Reagent Bottle
For Use With (Equipment) Chemicals in liquid or powder form
Height (English) 4 in.
Chemical Label Unlabeled
Closure Material GL45
Diameter (Metric) 3 cm
Closure Type Screw
Autoclavable Autoclavable
Show More Show Less