missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Class B Clear Glass Threaded Vials With Closures Packaged Separately
General-purpose vial
$134.67 - $269.59
Specifications
| Classification | B |
|---|---|
| Material (Liner) | Polyvinyl |
| Color | Clear |
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Catalog Number | Volume (Metric) | Height (Metric) | Diameter (Metric) Outer | Closure Size | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Volume (Metric) | Height (Metric) | Diameter (Metric) Outer | Closure Size | Price | Quantity & Availability | |||
|
0333921H
|
30 mL | 95 mm | 25 mm | 22-400 |
Pack of 144 Each for $194.97
Case of 4 Pack for $779.88
|
|
|||
|
0333921F
|
16 mL | 70 mm | 21 mm | 18-400 |
Pack of 144 Each for $179.81
Case of 8 Pack for $1,438.48
|
|
|||
|
0333921G
|
24 mL | 85 mm | 23 mm | 20-400 |
Pack of 144 Each for $190.97
Case of 6 Pack for $1,145.82
|
|
|||
|
0333921M
|
35.1 mL | 95 mm | 28 mm | 24-400 |
Pack of 144 Each for $266.48
Case of 3 Pack for $799.44
|
|
|||
|
0333921J
|
16 mL | 57 mm | 28 mm | 24-400 |
Pack of 144 Each for $233.97
Case of 3 Pack for $701.91
|
|
|||
|
0333921D
|
8 mL | 60 mm | 17 mm | 15-425 |
Pack of 144 Each for $141.10
Case of 12 Pack for $1,693.20
|
|
|||
|
0333921C
|
5.5 mL | 50 mm | 16 mm | 13-425 |
Pack of 144 Each for $136.00
Case of 16 Pack for $2,176.00
|
|
|||
|
0333921A
|
2 mL | 35 mm | 12 mm | 8-425 |
Pack of 144 Each for $166.03
Case of 16 Pack for $2,656.48
|
|
|||
|
0333921B
|
4 mL | 45 mm | 15 mm | 13-425 |
Pack of 144 Each for $134.67
Case of 16 Pack for $2,154.72
|
|
|||
|
0333921N
|
41 mL | 108 mm | 28 mm | 24-400 |
Pack of 144 Each for $269.59
Case of 3 Pack for $808.77
|
|
|||
|
0333921K
|
24 mL | 70 mm | 28 mm | 24-400 |
Pack of 144 Each for $251.62
Case of 3 Pack for $754.86
|
|
|||
|
0333921E
|
12 mL | 65 mm | 19 mm | 15-425 |
Pack of 144 Each for $161.23
Case of 8 Pack for $1,289.84
|
|
|||
Description
General-purpose vial
- Made from Type I, Class B borosilicate glass
- Black phenolic screw-top closure
- GPI thread finish
Specifications
| B | |
| Clear | |
| Non-sterile | |
| For general purpose | |
| Screw Cap (Closed-top) | |
| Vial |
| Polyvinyl | |
| Borosilicate Glass | |
| Single-use | |
| Phenolic | |
| ASTM E438, USP Type I |