missing translation for 'onlineSavingsMsg'
Learn More

Citronellol, 95%, Thermo Scientific™

Identifiants chimiques

CAS 106-22-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00002935
InChI Key QMVPMAAFGQKVCJ-UHFFFAOYSA-N
Synonym citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
PubChem CID 8842
ChEBI CHEBI:50462
IUPAC Name 3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC110460050
FDS Afficher les documents
Thermo Scientific Chemicals
110460050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC110461000
FDS Afficher les documents
Thermo Scientific Chemicals
110461000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC110465000
FDS Afficher les documents
Thermo Scientific Chemicals
110465000
500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

106-22-9
156.27
QMVPMAAFGQKVCJ-UHFFFAOYSA-N
8842
3,7-dimethyloct-6-en-1-ol
C10H20O
MFCD00002935
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
CHEBI:50462
CC(CCC=C(C)C)CCO

Spécifications

106-22-9
222.0°C
Authentic
Glass bottle
1.4550 to 1.4580
MFCD00002935
01, 451
15, 2329
Solubility in water: slightly soluble
CC(CCC=C(C)C)CCO
156.27
CHEBI:50462
95%
0.8540g/mL
79°C
94% min. (GC)
C10H20O
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
5mL
0.854
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
QMVPMAAFGQKVCJ-UHFFFAOYSA-N
3,7-dimethyloct-6-en-1-ol
8842
156.27
Citronellol, 95%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/ey

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-375-0

missing translation for 'rtecsNumber' : RH3400000

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only