missing translation for 'onlineSavingsMsg'
Learn More

Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol

Supplier:  Thermo Scientific Chemicals 04306630

 Afficher plus de versions de ce produit

Numéro de catalogue. AA4306630


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Chromium pieces
7440-47-3
Odorless
(metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
23976
99.99%
1 to 25mm (0.04 to 1 in.)
250 g
Broken Plate
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
[Cr]
52.00
CHEBI:28073

Identifiants chimiques

7440-47-3
52.00
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 231-157-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only