missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylvinylsilane, 97%

CAS: 1719-58-0 | C4H8ClSi | 119.64 g/mol

Supplier:  Thermo Scientific Chemicals L1619822

 Afficher plus de versions de ce produit

Numéro de catalogue. AAL1619822

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Store under dry inert gas.
Spécifications

Spécifications

82°C to 83°C
1719-58-0
C4H8ClSi
UN2985
−12°C (10°F)
Reacts with water.
C[Si](C)\C=C\Cl
119.64
120.66
1.416
0.883
Chlorodimethylvinylsilane
100 g
MFCD00018090
1737688
chlorodimethylvinylsilane, dimethylvinylchlorosilane, vinyldimethylchlorosilane, chloro dimethyl vinylsilane, silane, chloroethenyldimethyl, chlorodimethyl vinyl silane, unii-xja6n5221t, chloroethenyldimethyl-silane, chloro ethenyl dimethylsilane, chloro-dimethyl-ethenyl-silane
RABBDIBWJVOAPB-ONEGZZNKSA-N
[(1E)-2-chloroethenyl]dimethylsilyl
519368
97%
Moisture sensitive

Identifiants chimiques

1719-58-0
119.64
RABBDIBWJVOAPB-ONEGZZNKSA-N
519368
C4H8ClSi
MFCD00018090
chlorodimethylvinylsilane, dimethylvinylchlorosilane, vinyldimethylchlorosilane, chloro dimethyl vinylsilane, silane, chloroethenyldimethyl, chlorodimethyl vinyl silane, unii-xja6n5221t, chloroethenyldimethyl-silane, chloro ethenyl dimethylsilane, chloro-dimethyl-ethenyl-silane
C[Si](C)\C=C\Cl
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P374-P501c

H225-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 217-007-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only