Learn More
MilliporeSigma™ Chemicon™ Human Neural Stem Cell Characterization Kit
Catalog No. SCR060
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR060 1 Kit
1 options
Catalog No. SCR060 Supplier MilliporeSigma™ Supplier No. SCR060
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used in immunocytochemistry on human neural stem cells. MilliporeSigma™; Chemicon™ Cardiomyocyte Characterization Kit contains three molecular markers, Nestin, Sox 2 and Musashi that are frequently used to identify neural stem/progenitor cells.
  • Includes Mouse and rabbit immunoglobulins for assessment of background staining
  • Recommended that kit be used in conjunction with differentiation assays that demonstrate multipotentiality of starting cell population
Specifications
Product Type Stem Cell Characterization Kit
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • Mouse anti-Nestin, 50ug
  • Rabbit anti-Sox-2, 20ug
  • Rabbit anti-Musashi, 50ug
  • Mouse anti-bIII Tubulin, 50uL
  • Rabbit anti-GFAP, 50uL
  • Mouse anti-Oligodendrocyte marker O1, 20ug
  • Mouse IgM, 50ug
  • Mouse IgG, 100ug
  • Rabbit IgG, 100ug
When stored at the recommended storage conditions, components are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date.