missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cell Therapy Systems Hibernate™-E CTS™ Medium

Catalog No. A1370601
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1370601 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1370601 Supplier Gibco™ Supplier No. A1370601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell Therapy Systems Hibernate™-E CTS™ Medium

Cell Therapy Systems Hibernate™-E CTS™ media is a serum-free, nutrient basal media for the short-term maintenance of embryonic tissue in ambient CO2 conditions (Ref 1). Human tissues maintained in Hibernate™-E media include brain tumors (Ref 2), umbilical cords, and foreskins.

Quick and Easy Shipment of Human Tissues
Cell Therapy Systems Hibernate™-E CTS™ media can be used for shipment of human embryonic tissue as brain tumor, umbilical cord, and foreskin at ambient temperature using a simple 3 step process:

Collect tissue in a conical tube containing Hibernate medium (brain tumor cells require additional supplements).

  • Ship tissues in Hibernate™ media within 24 to 72 hours on cold packs.
  • Transfer received tissues into media of choice and initiate cultivation.
    Simplify Your Transition into Clinical Research
    The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ Cell Therapy Systems (CTS™) products deliver:
  • Reduced burden in qualifying reagents
  • Included appropriate certificates of analyses and origin
  • Convenient access to our Drug Master File (DMF)

    References:

  • Brewer, G.J. and Price, P.J. (1996).Viable cultured neurons in ambient carbon dioxide and hibernation storage for a month. Neuroreports 7: 1509-1512.
  • Brewer, G.J. and LeRoux, P.D.(2007). Human primary brain tumor cell growth inhibition in serum-free medium optimized for neuron survival. Brain Res. 1157:156-66.

  • Specifications

    Cell Type Neural Cells
    Classification Serum-free
    Form Liquid
    Product Type CTS Medium
    Serum Level Serum-free
    With Additives Phenol Red
    Product Line CTS, Hibernate
    Quantity 500 mL
    Shipping Condition Ambient
    Tested For Cytotoxicity, Endotoxin, Sterility, pH, Osmolality
    Content And Storage Store in refrigerator (2–8°C) and protect from light.

    Culture Type Mammalian Cell Culture
    Show More Show Less

    For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.