| Numéro d'enregistrement |
P27512 |
| Plage de l'essai |
3-1500 pg/mL |
| Sensibilité d’essai |
3 pg/mL |
| Plage d’étalonnage |
3 pg/mL to 1500 pg/mL |
| Contenu et stockage |
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Essai |
Solid-phase sandwich ELISA |
| Conjugué |
HRP |
| Format de la trousse d’immunodosage |
Sandwich ELISA Kit |
| Type de produit |
ELISA |
| Famille de protéines |
CD Proteins |
| Type d’échantillon |
Plasma, Serum, Supernatant |
| Spécificité |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, MIG, MIP- 1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. |
| Suffisant pour |
96 assays |
| Espèces cibles |
Mouse |
| À utiliser avec (application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| À utiliser avec (équipement) |
Absorbance Microplate Reader |
| Alias du gène |
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5; AI326936; Bp50; GP39; HIGM1; IGM; IMD3; T-BAM; TRAP; Tnfrsf5; p50 |
| ID du gène (Entrez) |
21939 |
| Symbole de gène |
CD40 |
| CV intra-essai |
<12%, <10% |
| Contenus des kits |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Étiquette ou colorant |
Biotin |
| Alias des protéines |
B-cell surface antigen CD40, Bp50, CD40L receptor |
| Quantité |
96 Tests |
| Statut réglementaire |
RUO |
| Disciplines de recherche |
Immunology |
| Volume d’échantillon |
Plasma 33 μL, Serum 33 μL, Supernatant 100 μL |
| Shipping condition (Condition d'expédition) |
Wet or Dry Ice |
| Conditions de stockage |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonyme |
RP23-428M13.1, AI326936, Bp50, GP39, HIGM1, IGM, IMD3, T-BAM, TRAP, Tnfrsf5, p50 |
| Cible |
CD40 |
| Durée du test |
1 hr 20 min |
| Durée totale de l'essai |
4 hr 45 min |
|
Afficher plus de résultats
Afficher moins de résultats
|
|