missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Canine TIMP-2 ELISA Kit
Description
Canine TIMP-2 quantitates canine TIMP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine TIMP-2.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Spécifications
Spécifications
| Numéro d'enregistrement | collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; Metalloproteinase inhibitor 2; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; Timp2; TIMP-2; TIMP-2 protein; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2 |
| Plage de l'essai | 20 to 5,000 pg/mL |
| Sensibilité d’essai | 20 pg/mL |
| Conjugué | HRP |
| Type de produit | ELISA |
| Type d’échantillon | Plasma, Serum, Supernatant |
| À utiliser avec (équipement) | Colorimetric Microplate Reader |
| ID du gène (Entrez) | 403633 |
| Symbole de gène | TIMP2 |
| CV intra-essai | <12%, <10 |
| Afficher plus de résultats |