missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Camptothecin

Cytotoxic antitumor agent that inhibits topoisomerase I | CAS: 7689-03-4 | C20H16N2O4

Supplier:  Thermo Scientific Chemicals J6252306

Quantity 5 g

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ6252306


Il en reste null

Description

Description

Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

3-4-7689
(S)-(+)-Camptothecin

Spécifications

Yellow
3-4-7689
348.35g/mol
260°C
2°C to 8°C
Powder
5 g
C20H16N2O4
(S)-(+)-Camptothecin
Apoptosis Reagent
Cytotoxic antitumor agent that inhibits topoisomerase I
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions