missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Carboxymethylcellulose, Sodium Salt, Low Viscosity

Substrate for cellulase

Supplier:  MilliporeSigma™ 217277250GM

 Afficher plus de versions de ce produit

Numéro de catalogue. 217277250GM


Il en reste null

Description

Description

  • Substrate for cellulase
  • Viscosity: (2%, H2O, 25°C): 25 - 75 mPas
Spécifications

Identifiants chimiques

White to tan
Store at +15°C to +30°C
Free-flowing Solid

Spécifications

White to tan
Store at +15°C to +30°C
250 g
Free-flowing Solid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions