missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Aquacide II

Highly viscous salt useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing

$206.00 - $868.00

Specifications

CAS 9004-32-4
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 500000g/mol
Form Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
178511KG
View Documents Promotion Details
MilliporeSigma™
178511KG
1 kg
Each for $206.00
Only null left
 
178515KG
View Documents Promotion Details
MilliporeSigma™
178515KG
5 kg
Each for $868.00
Only null left
 
Description

Description

  • Sodium salt of carboxymethylcellulose
  • High viscosity
  • Useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing by using either a 2% syrup or the Aquacide solid
  • Neither denature proteins nor change the pH during concentration
  • At room temperature 1g of Aquacide absorbs about 5mL of water from solution in 1h
Specifications

Specifications

9004-32-4
Store at +15°C to +30°C
Solid
White
500000g/mol
Videos
SDS
Documents

Documents