missing translation for 'onlineSavingsMsg'
Learn More

Wanzl Bulk Shelving Add On Unit

Bulk shelving systems from Wanzl provides maximum utilization and high load capacity. Our shelves are built for strength and durability and hold up to 900 lb. per shelf. Starter and add on units are available, as well as a variety of accessories for customization.

Supplier:  Wanzl TBCR367A24

 Afficher plus de versions de ce produit

Numéro de catalogue. 11222131


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Tool-free installation - adjustable in 2 in. increments.
  • Black powder-coated with e-coat - corrosion, chemical and chip resistant.
  • 10 Year warranty against rust/corrosion and 1 year defective material/workmanship warranty.
Spécifications

Spécifications

36 in.
Add on Unit
4
Steel
24 x 36 x 78 in.
Shelving
Chrome
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions