missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Bulk pack of amber wide-mouth HDPE reagent bottles with polypropylene caps are UV-protective, leak-resistant, and chemically durable, perfect for storing and transporting light-sensitive liquids in high-volume lab or industrial use.
$1,036.64 - $1,954.57
Spécifications
| Color | Amber |
|---|---|
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Numéro de catalogue | Numéro du manufacturier. | Capacity (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Capacity (Metric) | Prix | Quantity & Availability | |||||
|
S000141
|
Eisco Labs
CH5540PK500 |
125 mL |
paquet de 500 chaque for $1,954.57
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
S000142
|
Eisco Labs
CH5542PK125 |
500 mL |
paquet de 125 chaque for $1,036.64
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Bulk pack for lab needs: Bulk pack of amber wide-mouth HDPE reagent bottles with polypropylene caps, ideal for labs needing large quantities for light-sensitive storage and transport.
- UV light shielding: Amber-colored bottles block harmful UV rays, making them ideal for storing and handling light-sensitive substances in laboratories and industrial settings.
- Leak-proof and reliable: Designed with a secure seal, ensuring safe containment of reagents and chemicals during storage and transport.
- Chemical resistance: Made from HDPE, these bottles are durable and resistant to a wide range of chemicals, suitable for various laboratory, industrial, and research applications.
- Designed for convenience: Features a wide neck for easy filling and pouring, and lightweight construction for hassle-free transport.
Spécifications
| Amber | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| HDPE | |
| Yes | |
| Wide | |
| Not Autoclavable |