missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Bulk Pack of Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000142
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000142 500 mL
S000141 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000142 Supplier Eisco Labs Supplier No. CH5542PK125

Bulk pack of amber wide-mouth HDPE reagent bottles with polypropylene caps are UV-protective, leak-resistant, and chemically durable, perfect for storing and transporting light-sensitive liquids in high-volume lab or industrial use.

  • Bulk pack for lab needs: Bulk pack of amber wide-mouth HDPE reagent bottles with polypropylene caps, ideal for labs needing large quantities for light-sensitive storage and transport.
  • UV light shielding: Amber-colored bottles block harmful UV rays, making them ideal for storing and handling light-sensitive substances in laboratories and industrial settings.
  • Leak-proof and reliable: Designed with a secure seal, ensuring safe containment of reagents and chemicals during storage and transport.
  • Chemical resistance: Made from HDPE, these bottles are durable and resistant to a wide range of chemicals, suitable for various laboratory, industrial, and research applications.
  • Designed for convenience: Features a wide neck for easy filling and pouring, and lightweight construction for hassle-free transport.

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Amber
Diameter (Metric) Inner Neck 43.7 mm
Material HDPE
No. per Pack 125 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 167 mm
Product Type Reagent Bottle
Diameter (Metric) 73.7 mm
Autoclavable Not Autoclavable
Show More Show Less