missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 1.00 (Certified), Fisher Chemical

Manufacturer: Fisher Chemical BPSB140500
Specifications
Buffer Solution, pH 1.01 | |
MFCD00146206 | |
Liquid | |
Poly Bottle | |
1.0g/cm³ | |
Traceable to NIST. | |
0°C | |
Undesignated |
7447-40-7,7647-01-0,50-00-0,67-56-1,7732-18-5 | |
In Pass Test | |
Certified | |
Pass Test | |
Potassium Chloride,Hydrochloric Acid | |
100°C | |
1.00 | |
500mL |