missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical
Spécifications
Spécifications
| Chemical Name or Material | Buffer Solution |
| Melting Point | ∼ 0°C |
| Color | Colorless |
| For Use With (Application) | Ion Selective Analysis |
| pH | 2.98 to 3.98 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Packaging | Amber Glass Bottle |
| Grade | Chemical Solution |