accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Electrochemistry Buffers
SB142500
Print
Buffer Concentrate, pH 11.00 (Certified), Fisher Chemical
Supplier:
Fisher Chemical
SB142500
Catalog No.
SB142-500
Sign In
or
Register
to check your price and availability.
$315.50
/
Each
$1,893.00
/
Case of 6 Each
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Quantity
Only null left
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Buffer Concentrate, pH 11.01
CAS
56-40-6
,
7647-14-5
,
1310-73-2
,
139-33-3
,
7732-18-5
Physical Form
Liquid
Identification
Pass Test
Packaging
Poly Bottle
Vapor Pressure
14mmHg
Melting Point
0°C
Color
Undesignated
Quantity
500 mL
MDL Number
MFCD00163176
Traceability to NIST
Traceable to NIST.
Grade
Certified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions