Learn More
BRANDTECH™ VITLAB™ High-density Polyethylene Funnels
VITLAB™ large high density polyethylene funnels are suitable for transfers of large volumes of liquid.
Supplier:  BRANDTECH™ VITLAB™ 42294
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-378-997
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Integrated handle facilitates use
Spécifications
Spécifications
400mm
42mm
365 mm
Filling Funnel (Liquid)
12,500 mL
HDPE
Handle
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS