Learn More
Brady™ ToughStripe™ Floor Marking Prespaced Marks
ToughStripe™ Floor Marking Prespaced Marks are made of B-514 polyester material that is ideal for use in marking aisles, passageways, storage locations, and point out safety equipment and exits.
$112.61 - $124.31
Specifications
Certifications/Compliance ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Adhesive Type Rubber, Permanent
Coating None
Finish Gloss
Format Tape
View More Specs
Products 7
Catalog Number Mfr. No. Color Shape Price Quantity  
Catalog Number Mfr. No. Color Shape Price Quantity  
19150842
View Documents
Brady™
104556
Yellow Rectangle
Each for $112.61
Only null left
19150843
View Documents
Brady™
104520
Green Round
Each for $124.31
Only null left
19150844
View Documents
Brady™
104521
Red Round
Each for $124.31
Only null left
19150846
View Documents
Brady™
104523
Yellow Round
Each for $124.31
Only null left
19150845
View Documents
Brady™
104522
White Round
Each for $124.31
Only null left
19150847
View Documents
Brady™
104524
Blue Round
Each for $124.31
Only null left
19150848
View Documents
Brady™
104525
Black Round
Each for $124.31
Only null left
Description
Description
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Tape removes cleanly without any unsightly adhesive residue or damage to the floor
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.36
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
None
Tape
Single-side Printable
Label Tape
115 Gardner Units
1 Roll
-18°C to +54°C
0.201 mm
Rubber, Permanent
Gloss
Polyester
None
Non-sterile
-18°C
0°F to 130°F
0.008 in.
100 ft.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'