Learn More
Brady™ ToughStripe™ Floor Marking Prespaced Marks
ToughStripe™ Floor Marking Prespaced Marks are made of B-514 polyester material that is ideal for use in marking aisles, passageways, storage locations, and point out safety equipment and exits.
Supplier:  Brady™ 104523
 View more versions of this product
Catalog No. 19150846
Only null left
Explore available promotions
Description
Description
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Tape removes cleanly without any unsightly adhesive residue or damage to the floor
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.36
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Yellow
Tape
Single-side Printable
Label Tape
For floor dot, mark aisles, passageways, storage locations, point out safety equipment, exits
-18°C
1 Roll
0°F to 130°F
0.008 in.
88.9 x 88.9 mm
3.5 in.
3.5 in.
TS-88.90-514-PREDIA-YL-RL
ToughStripe Prespaced Shapes
None
Gloss
Polyester
None
Non-sterile
115 Gardner Units
80 Pieces/Roll
Round
-18°C to +54°C
0.201 mm
100 ft.
88.9 mm
8.89 cm
Product Suggestions
Product Suggestions