Learn More
Brady™ ToughStripe™ Floor Marking Prespaced Marks
ToughStripe™ Floor Marking Prespaced Marks are made of B-514 polyester material that is ideal for use in marking aisles, passageways, storage locations, and point out safety equipment and exits.
Supplier:  Brady™ 104556
 View more versions of this product
Catalog No. 19150842
Only null left
Explore available promotions
Description
Description
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Tape removes cleanly without any unsightly adhesive residue or damage to the floor
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.36
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Yellow
Tape
Single-side Printable
Label Tape
For floor dash, mark aisles, passageways, storage locations, point out safety equipment, exits
-18°C
1 Roll
0°F to 130°F
0.008 in.
203.20 x 50.8 mm
30.48 m
50.8 mm
203.20 mm
ToughStripe Prespaced shapes
None
Gloss
Polyester
None
Non-sterile
115 Gardner Units
65 Pieces/Roll
Rectangle
-18°C to +54°C
0.201 mm
100 ft.
2 in.
8 in.
TS-50.80-514-PREDASH-YL-RL
Product Suggestions
Product Suggestions